Please login to your online account to display your discounted pricing

anti-SSEA-4, NL493, Clone: MC-813-70, R&D Systems

$452.99 - $585.69

Specifications

Antigen SSEA-4
Clone MC-813-70
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NLLC1435G

 
r & d systems
RDSNLLC1435G
NL493 0.5mL Each for $585.69

NL1435G

 
r & d systems
RDSNL1435G
NL493 0.5mL Each for $452.99
Description & Specifications

Specifications

Antigen SSEA-4
Clone MC-813-70
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen 2120Ep human embryonal carcinoma cell line.
Isotype IgG3
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Immunocytochemistry