Please login to your online account to display your discounted pricing

anti-SSEA-4, NL493, Clone: MC-813-70, R&D Systems


Manufacturer: r & d systems  RDSNL1435G

 View more versions of this product

Catalog No. NL1435G

  • / Each
 

Description & Specifications

Specifications

Isotype IgG3
Format Purified
Antigen SSEA-4
Immunogen 2120Ep human embryonal carcinoma cell line.
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Conjugate NL493
Primary or Secondary Primary
Quantity 0.5mL
Purification Method Protein A or G purified from hybridoma culture supernatant
Clone MC-813-70