Please login to your online account to display your discounted pricing

anti-SSEA-1, NL557, Clone: MC-480, R&D Systems


Manufacturer: r & d systems  RDSNLLC2155R

 View more versions of this product

Catalog No. NLLC2155R

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Immunogen F9 mouse teratocarcinoma stem cells.
Isotype IgM
Antigen SSEA-1
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Primary or Secondary Primary
Conjugate NL557
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Clone MC-480
Regulatory Status RUO
Purification Method IgM-specific Affinity-purified from hybridoma culture supernatant