Please login to your online account to display your discounted pricing

anti-SSEA-1, NL557, Clone: MC-480, R&D Systems

$452.99 - $585.69

Specifications

Antigen SSEA-1
Clone MC-480
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NLLC2155R

 
r & d systems
RDSNLLC2155R
NL557 0.5mL Each for $585.69

NL2155R

 
r & d systems
RDSNL2155R
NL557 0.5mL Each for $452.99
Description & Specifications

Specifications

Antigen SSEA-1
Clone MC-480
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Host Species Mouse
Regulatory Status RUO
Immunogen F9 mouse teratocarcinoma stem cells.
Isotype IgM
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method IgM-specific Affinity-purified from hybridoma culture supernatant

Applications:

Immunocytochemistry