Please login to your online account to display your discounted pricing

anti-SSEA-1, NL557, Clone: MC-480, R&D Systems


Manufacturer: r & d systems  RDSNL2155R

 View more versions of this product

Catalog No. NL2155R

  • / Each
 

Description & Specifications

Specifications

Antigen SSEA-1
Host Species Mouse
Format Affinity Purified
Immunogen F9 mouse teratocarcinoma stem cells.
Isotype IgM
Regulatory Status RUO
Clone MC-480
Cross Reactivity Human
Cross Reactivity Mouse
Purification Method IgM-specific Affinity-purified from hybridoma culture supernatant
Quantity 0.5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate NL557