Please login to your online account to display your discounted pricing

anti-SSEA-1, NL493, Clone: MC-480, R&D Systems


Manufacturer: r & d systems  RDSNLLC2155G

 View more versions of this product

Catalog No. NLLC2155G

  • / Each
 

Description & Specifications

Specifications

Antigen SSEA-1
Host Species Mouse
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgM
Quantity 0.5mL
Format Affinity Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone MC-480
Conjugate NL493
Immunogen F9 mouse teratocarcinoma stem cells.
Purification Method IgM-specific Affinity-purified from hybridoma culture supernatant