Please login to your online account to display your discounted pricing

anti-SSEA-1, APC, Clone: MC-480, R&D Systems

$432.57 - $432.57

Specifications

Clone MC-480
Host Species Mouse
Isotype IgM
Cross Reactivity Human
Cross Reactivity Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB2155A100

 
r & d systems
FAB2155A100
APC 100 Tests Each for $432.57
Description & Specifications

Specifications

Clone MC-480
Host Species Mouse
Isotype IgM
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen F9 mouse teratocarcinoma stem cells
Antigen SSEA-1
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Test Specificity Detects human and mouse SSEA-1.
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Purification Method IgM-specific Affinity-purified from hybridoma culture supernatant