Please login to your online account to display your discounted pricing

anti-SSEA-1, APC, Clone: MC-480, R&D Systems


Manufacturer: r & d systems  FAB2155A100

 View more versions of this product

Catalog No. FAB2155A100

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Host Species Mouse
Regulatory Status RUO
Quantity 100 Tests
Applications Flow Cytometry
Immunogen F9 mouse teratocarcinoma stem cells
Isotype IgM
Format Affinity Purified
Clone MC-480
Antigen SSEA-1
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Primary or Secondary Primary
Purification Method IgM-specific Affinity-purified from hybridoma culture supernatant
Monoclonal or Polyclonal Monoclonal
Test Specificity Detects human and mouse SSEA-1.
Conjugate APC