missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
single-stranded DNA binding protein 2 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SSBP2 protein.
Supplier: Abnova Corporation H00023635B01
Description
Sequence: MYGKGKSNSSAVPSDSQAREKLALYVYEYLLHVGAQKSAQTFLSEIRWEKNITLGEPPGFLHSWWCVFWDLYCAAPERRETCEHSSEAKAFHDYSAAAAPSPVLGNIPPGDGMPVGPVPPGFFQPFMSPRYPGGPRPPLRIPNQALGGVPGSQPLLPSGMDPTRQQGHPNMGGPMQRMTPPRGMVPLGPQNYGGAMRPPLNALGGPGMPGMNMGPGGGRPWPNPTNANSIPYSSASPGNYVGPPGGGGPPGTPIMPSPADSTNSGDNMYTLMNAVPPGPNRPNFPMGPGSDGPMGGLGGMESHHMNGSLGSGDMDSISKNSPNNMSLSNQPGTPRDDGEMGGNFLNPFQSESYSPSMTMSVSpecifications
single-stranded DNA binding protein 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human SSBP2 protein. | |
SSBP2 | |
DKFZp686F03273/HSPC116 | |
Mouse | |
50 μL | |
Yes | |
23635 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence,Western Blot | |
Unconjugated | |
No additive | |
BC017020 | |
SSBP2 | |
SSBP2 (AAH17020, 1 a.a. ∼ 361 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |