Please login to your online account to display your discounted pricing

anti-SREBP1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a portion of the human SREBP1 protein sequence (between residues 700-800). [UniProt No. P36956]
Antigen SREBP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10060545

 
novus biologicals canada
NB10060545
Unlabeled 0.1mL Each for $329.00

NB10060545T

 
novus biologicals canada
NB10060545SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a portion of the human SREBP1 protein sequence (between residues 700-800). [UniProt No. P36956]
Antigen SREBP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human, mouse, rat, and hamster. Predicted to react with porcine based on 100% sequence homology. Immunogen sequence has 94% homology to bovine, 88% homology to Zebrafish and 70% homology to chicken.
Gene Accession No. P36956
Gene ID (Entrez) 6720
Formulation Tris-glycine, 150mM NaCl
Concentration 1.1mg/mL