Please login to your online account to display your discounted pricing

anti-SREBP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10060545SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10060545T

  • / Each
 

Description & Specifications

Specifications

Antigen SREBP1
Applications Western Blot
Concentration 1.1mg/mL
Conjugate Unlabeled
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P36956
Host Species Rabbit
Immunogen A synthetic peptide made to a portion of the human SREBP1 protein sequence (between residues 700-800). [UniProt No. P36956]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6720
Test Specificity Human, mouse, rat, and hamster. Predicted to react with porcine based on 100% sequence homology. Immunogen sequence has 94% homology to bovine, 88% homology to Zebrafish and 70% homology to chicken.
Target Species Hamster
Target Species Human
Target Species Murine
Target Species Rat