Please login to your online account to display your discounted pricing

anti-SREBP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10060545

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10060545

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 6720
Monoclonal or Polyclonal Polyclonal
Formulation Tris-glycine, 150mM NaCl
Primary or Secondary Primary
Immunogen A synthetic peptide made to a portion of the human SREBP1 protein sequence (between residues 700-800). [UniProt No. P36956]
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Test Specificity Human, mouse, rat, and hamster. Predicted to react with porcine based on 100% sequence homology. Immunogen sequence has 94% homology to bovine, 88% homology to Zebrafish and 70% homology to chicken.
Gene Accession No. P36956
Concentration 1.1mg/mL
Conjugate Unlabeled
Format Affinity Purified
Applications Western Blotting
Quantity 0.1mL
Antigen SREBP1
Host Species Rabbit
Regulatory Status RUO