Please login to your online account to display your discounted pricing

anti-SRC Clone: 5D10C4

SRC Monoclonal antibody MA5-15412 from Thermo Scientific™ specifically detects SRC in Human samples, and it is validated for Western Blot

$506.05 - $506.05

Specifications

Antigen SRC
Clone 5D10C4
Applications Western Blotting
Applications Western Blotting
Classification Monoclonal
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

PIMA515412

 
pierce biotechnology inc
MA515412
Each for $506.05
Description & Specifications

Specifications

Antigen SRC
Clone 5D10C4
Applications Western Blotting
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Ascites
Host Species Mouse
Quantity 100μL
Regulatory Status RUO
Type Primary
Formulation 100μL unpurified antibody in ascites and 0.03% sodium azide
Gene Symbols SRC
Immunogen Purified recombinant fragment of SRC (aa10-193) expressed in E. Coli.
Isotype IgG2a
Storage Requirements avoiding freeze/thaw cycles.

SRC Monoclonal antibody MA5-15412 from Thermo Scientific™ specifically detects SRC in Human samples, and it is validated for Western Blot

This gene is highly similar to the v-src gene of Rous sarcoma virus. This proto-oncogene may play a role in the regulation of embryonic development and cell growth. The protein encoded by this gene is a tyrosine-protein kinase whose activity can be inhibited by phosphorylation by c-SRC kinase. Mutations in this gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same protein have been found for this gene.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG2a
Clone: 5D10C4
Immunogen: Purified recombinant fragment of SRC (aa10-193) expressed in E. Coli.

Formulation: 100μL unpurified Antibody in ascites, with 0.03% sodium azide;
Synonyms: ASV; SRC1; c-SRC; p60-Src; SRC
Uniprot Accession: P12931
Gene Symbol: SRC
Entrez GeneID: 6714
MA5-15412 detects SRC in Human samples. MA5-15412 has been successfully used in Western Blot procedures. MA5-15412 The immunogen is Purified recombinant fragment of SRC (aa10-193) expressed in E. Coli.
.