Please login to your online account to display your discounted pricing

anti-SR1, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 45-76 of human Sorcin was used as immunogen
Antigen SR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224479SS

 
novus biologicals canada
NBP224479SS
Unlabeled 0.025mg Each for $109.00

NBP224479

 
novus biologicals canada
NBP224479
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 45-76 of human Sorcin was used as immunogen
Antigen SR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b and CRA_c), cow, rat and mouse, 93% in xenopus, 87% in opossum and 81% homologous in chicken.
Gene Accession No. Q7T365
Gene ID (Entrez) 6717
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL