Please login to your online account to display your discounted pricing

anti-SR-BI/SR-BII, Polyclonal, Novus Biologicals™

$144.63 - $329.00

Specifications

Host Species Rabbit
Immunogen A peptide from the extracellular domain (residues 230-380) of Scavenger Receptor-BI/BII that was expressed as two tandem copies in bacteria using the pET system.
Antigen SR-BI/SR-BII
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB400134

 
novus biologicals canada
NB400134
Unlabeled 0.1mL Each for $329.00

NB400134SS

 
novus biologicals canada
NB400134SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A peptide from the extracellular domain (residues 230-380) of Scavenger Receptor-BI/BII that was expressed as two tandem copies in bacteria using the pET system.
Antigen SR-BI/SR-BII
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Mouse. Previous lots have been shown to react with rat and human as well.
Gene ID (Entrez) 949