Please login to your online account to display your discounted pricing

SR-BI Polyclonal Antibody, Invitrogen™


SR-BI Antibody, PA1-16803, from Invitrogen™. Species Reactivity: Hamster, Human, Mink, Mouse, Rat, Rodent; Applications: FACS, ICC, IF, IHC (Paraffin), WB.

Manufacturer: pierce biotechnology inc  PA116803

Catalog No. APPA6803

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL purified antibody (3mg/mL) in tris citrate and 0.09% sodium azide; pH 7-8
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mink
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Rodent
Isotype IgG
Storage Requirements Store at 4° C
Gene Accession No. P97943, Q61009, Q8WTV0
Gene Symbols SCARB1
Gene Alias SCARB1, SR-B1, SR-BI, Scavenger Receptor-BI, Scavenger Receptor-B1
Gene ID (Entrez) 20778, 25073, 949
Species Reactivity Hamster, Human, Mink, Mouse, Rat, Rodent
Quantity 100μL
Format Purified
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Antigen SR-BI
Host Species Goat
Immunogen Synthetic peptide corresponding to the C-terminal residues 400-509 of mouse SR-BI.

SR-BI Antibody, PA1-16803, from Invitrogen. Species Reactivity: Hamster, Human, Mink, Mouse, Rat, Rodent; Applications: Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot.

High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. The SR-BI binds HDLs and mediates selective uptake of HDL cholesteryl ester. SR-BI binds HDL with high affinity, is expressed primarily in liver and nonplacental steroidgenic tissues, and mediates selective cholesterol uptake by a distinct mechanism. In mice, it seems that SR-BI plays a key role in determining the levels of plasma lipoprotein cholesterol and the accumulation of cholesterol stores in the adrenal gland.
Host Species: Goat
Cross Reactivity: Hamster, Human, Mink, Mouse, Rat, Rodent
Isotype: IgG
Immunogen: Synthetic peptide corresponding to the C-terminal residues 400-509 of mouse SR-BI.
Formulation: 100μL purified antibody (3mg/mL) in tris citrate and 0.09% sodium azide; pH 7-8
Synonyms: SCARB1, SR-B1, SR-BI, Scavenger Receptor-BI, Scavenger Receptor-B1
Uniprot Accession: P97943, Q61009, Q8WTV0
Entrez GeneID: 20778, 25073, 949
Gene Symbol: SCARB1
Suggested positive control: mouse liver lysate or protein.
References:
Test: WB, Species: Hm, Reference: Omega 3 fatty acids promote macrophage reverse cholesterol transport in hamster fed high fat diet. (2013) PLoS One, Vol: 8, 4, e61109, Kasbi Chadli F, Nazih H, Krempf M, Nguyen P, Ouguerram K; Pubmed ID: 23613796