Please login to your online account to display your discounted pricing

anti-SR-BI, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB400131SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB400131SS

  • / Each
 

Description & Specifications

Specifications

Antigen SR-BI
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 3mg/mL
Conjugate Unlabeled
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Mustelid
Cross Reactivity Rat
Cross Reactivity Squirrel
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Host Species Goat
Immunogen A synthetic peptide made to the C-terminal region of mouse SR-BI (between residues 400-509). [UniProt No. Q61009]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 949
Test Specificity Mouse, mink, rat, hamster and human. Customers have also reported positive detection in cytosolic fractions from 13-lined ground squirrel liver samples, with an unidentified non-specific band at ~60 kDa.
Target Species Hamster
Target Species Human
Target Species Murine
Target Species Mustelid
Target Species Rat
Target Species Squirrel