Please login to your online account to display your discounted pricing

anti-SR-BI, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB400131

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB400131

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Concentration 3mg/mL
Host Species Goat
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Mustelid
Cross Reactivity Rat
Cross Reactivity Squirrel
Conjugate Unlabeled
Gene ID (Entrez) 949
Test Specificity Mouse, mink, rat, hamster and human. Customers have also reported positive detection in cytosolic fractions from 13-lined ground squirrel liver samples, with an unidentified non-specific band at ~60 kDa.
Formulation Tris-citrate/phosphate, pH 7-8
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Affinity Purified
Antigen SR-BI
Quantity 0.1mL
Primary or Secondary Primary
Immunogen A synthetic peptide made to the C-terminal region of mouse SR-BI (between residues 400-509). [UniProt No. Q61009]