Please login to your online account to display your discounted pricing

anti-SR-BI, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Goat
Immunogen A synthetic peptide made to the C-terminal region of mouse SR-BI (between residues 400-509). [UniProt No. Q61009]
Antigen SR-BI
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB400131

 
novus biologicals canada
NB400131
Unlabeled 0.1mL Each for $329.00

NB400131SS

 
novus biologicals canada
NB400131SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Goat
Immunogen A synthetic peptide made to the C-terminal region of mouse SR-BI (between residues 400-509). [UniProt No. Q61009]
Antigen SR-BI
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Mouse, mink, rat, hamster and human. Customers have also reported positive detection in cytosolic fractions from 13-lined ground squirrel liver samples, with an unidentified non-specific band at ~60 kDa.
Gene ID (Entrez) 949
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 3mg/mL