missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SR-AI/MSR, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Macrophage Scavenger Receptor I (within residues 400-450). [Swiss-Prot No. P21757]
Antigen SR-AI/MSR
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP100092SS Novus Biologicals Canada
NBP100092SS
0.025mL Each for $139.00
NBP100092 Novus Biologicals Canada
NBP100092
0.1mL Each for $362.00
Specifications Description & Specifications

Specifications

Rabbit
SR-AI/MSR
Affinity Purified
RUO
Reacts with human and mouse. 100% sequence identity with horse and dog, 93% identity with monkey and bovine, 92% identity with rat.
4481
1.19mg/mL
Synthetic peptide made to an internal portion of human Macrophage Scavenger Receptor I (within residues 400-450). [Swiss-Prot No. P21757]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Western Blot
P21757
PBS, 30% glycerol