Please login to your online account to display your discounted pricing

anti-SR-AI/MSR, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Macrophage Scavenger Receptor I (between residues 300-350). [UniProt P21757]
Antigen SR-AI/MSR
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB400159

 
novus biologicals canada
NB400159
Unlabeled 0.1mL Each for $309.05

NB400159SS

 
novus biologicals canada
NB400159SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Macrophage Scavenger Receptor I (between residues 300-350). [UniProt P21757]
Antigen SR-AI/MSR
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human and monkey. There have been mixed results (positive and negative) with mouse.
Gene Accession No. P21757
Gene ID (Entrez) 4481
Formulation Tris-glycine, 150mM NaCl
Concentration 0.97mg/mL