Please login to your online account to display your discounted pricing

anti-SR-AI/MSR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP100093

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP100093

  • / Each
 

Description & Specifications

Specifications

Test Specificity Reacts with human and mouse. 100% sequence identity with horse and dog, 93% identity with monkey and bovine, 92% identity with rat.
Conjugate Unlabeled
Primary or Secondary Primary
Formulation PBS, 30% glycerol
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Gene ID (Entrez) 4481
Gene Accession No. P21757
Immunogen Synthetic peptide made to a cyclized peptide from human Macrophage Scavenger Receptor I (within residues 400-450). [Swiss-Prot No. P21757]
Antigen SR-AI/MSR
Format Affinity Purified