missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Spi-B, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 110-160 of human Spi-B was used as the immunogen.
Antigen Spi-B
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224625 Novus Biologicals Canada
NBP224625
0.1mg Each for $255.80
NBP224625SS Novus Biologicals Canada
NBP224625SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Spi-B
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, cow, chimpanzee, rat, mouse, and dog, 94% homologous in horse and 72% homologous in Xenopus.
6689
0.5mg/mL
A portion of amino acids 110-160 of human Spi-B was used as the immunogen.
Polyclonal
Primary
Western Blot
Q01892
100μg in 200μL PBS containing 0.05% BSA