Please login to your online account to display your discounted pricing

anti-Spi-B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224625SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224625SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 6689
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q01892
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Host Species Rabbit
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cow, chimpanzee, rat, mouse, and dog, 94% homologous in horse and 72% homologous in Xenopus.
Concentration 0.5mg/mL
Antigen Spi-B
Immunogen A portion of amino acids 110-160 of human Spi-B was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Format Purified