Please login to your online account to display your discounted pricing

anti-Sphingosine Kinase 1/SPHK1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP122974SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP122974SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q2M3C7
Primary or Secondary Primary
Conjugate Unlabeled
Format Affinity Purified
Concentration 1mg/mL
Host Species Rabbit
Gene ID (Entrez) 8877
Formulation Tris-citrate/phosphate buffer, pH 7 to 8
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Cross Reactivity Human
Antigen Sphingosine Kinase 1/SPHK1
Immunogen sphingosine kinase 1
Quantity 0.025mL
Applications Immunoprecipitation
Applications Western Blotting