Please login to your online account to display your discounted pricing

anti-Spectrin beta 3, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a C-terminal region between residue 2340 and 2390 of Spectrin beta 3 using the numbering given in entry NP_008877.1
Antigen Spectrin beta 3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11058346

 
novus biologicals canada
NB11058346
Unlabeled 0.1mL Each for $362.00

NB11058346T

 
novus biologicals canada
NB11058346SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a C-terminal region between residue 2340 and 2390 of Spectrin beta 3 using the numbering given in entry NP_008877.1
Antigen Spectrin beta 3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. O15020
Gene ID (Entrez) 6712
Formulation TBS with 0.1% BSA.
Concentration 0.2mg/mL