missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SPARC, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-210 of human SPARC was used as the immunogen for the antibody. (aa179-197)
Antigen SPARC
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227371 Novus Biologicals Canada
NBP227371
0.1mg Each for $255.80
NBP227371SS Novus Biologicals Canada
NBP227371SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
SPARC
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human and monkey and 95% homolgous in mouse and rat.
6678
0.5mg/mL
A portion of amino acids 150-210 of human SPARC was used as the immunogen for the antibody. (aa179-197)
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
P09486
0.1mg SPARC antibody in 0.2mL PBS containing 0.05% BSA.