Please login to your online account to display your discounted pricing

anti-SPARC, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227371

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227371

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation 0.1mg SPARC antibody in 0.2mL PBS containing 0.05% BSA.
Monoclonal or Polyclonal Polyclonal
Gene Accession No. P09486
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Quantity 0.1mg
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and monkey and 95% homolgous in mouse and rat.
Gene ID (Entrez) 6678
Host Species Rabbit
Antigen SPARC
Immunogen A portion of amino acids 150-210 of human SPARC was used as the immunogen for the antibody. (aa179-197)
Regulatory Status RUO