Please login to your online account to display your discounted pricing

anti-SOX2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227394

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227394

  • / Each
 

Description & Specifications

Specifications

Antigen SOX2
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS, 0.05% BSA.
Gene Accession No. P48431
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SOX2 protein (between residues 50-100) [UniProt P48431]
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6657
Target Species Human
Target Species Murine