missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SOX2, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SOX2 protein (between residues 50-100) [UniProt P48431]
Antigen SOX2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227394 Novus Biologicals Canada
NBP227394
0.1mg Each for $257.41
NBP227394SS Novus Biologicals Canada
NBP227394SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
SOX2
Purified
RUO
P48431
PBS, 0.05% BSA.
A synthetic peptide made to an internal portion of the human SOX2 protein (between residues 50-100) [UniProt P48431]
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
6657
0.5mg/mL