Please login to your online account to display your discounted pricing

anti-SOX17, Polyclonal, Novus Biologicals™

$109.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 70-120 of human SOX17 was used as the immunogen for the antibody.
Antigen SOX17
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224568

 
novus biologicals canada
NBP224568
Unlabeled 0.1mg Each for $362.00

NBP224568SS

 
novus biologicals canada
NBP224568SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 70-120 of human SOX17 was used as the immunogen for the antibody.
Antigen SOX17
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used is 100% homologous in human and 69% homologous in bovine.
Gene Accession No. Q9H6I2
Gene ID (Entrez) 64321
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL