Please login to your online account to display your discounted pricing

anti-SOX11, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A partial recombinant portion of human SOX11 (between residues 50-300) [UniProt P35716]
Antigen SOX11
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP231371

 
novus biologicals canada
NBP231371
Unlabeled 0.1mg Each for $307.00

NBP231371SS

 
novus biologicals canada
NBP231371SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A partial recombinant portion of human SOX11 (between residues 50-300) [UniProt P35716]
Antigen SOX11
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human. Immunogen sequence similarity with other species : Mouse (82%), Rat (82%), Chicken (88%), Xenopus (84%).
Gene Accession No. P35716
Gene ID (Entrez) 6664
Formulation PBS
Concentration 1.0mg/mL