Please login to your online account to display your discounted pricing

anti-SOX1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen 196 RAVHMkEYPDYkYRPRRk 213
Antigen SOX1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227189SS

 
novus biologicals canada
NBP227189SS
Unlabeled 0.025mg Each for $109.00

NBP227189

 
novus biologicals canada
NBP227189
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 196 RAVHMkEYPDYkYRPRRk 213
Antigen SOX1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acidsequence used as immunogen shares significant homology with SOX1, 2, 3, 7, 11, 12, 14, 15, 20, and 21. Dog, Human, New World monkey, Mouse, Opossum, Rat, Xenopus
Gene ID (Entrez) 6656
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL