Please login to your online account to display your discounted pricing

anti-SOX1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227189SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227189SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA.
Primary or Secondary Primary
Test Specificity The amino acidsequence used as immunogen shares significant homology with SOX1, 2, 3, 7, 11, 12, 14, 15, 20, and 21. Dog, Human, New World monkey, Mouse, Opossum, Rat, Xenopus
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Xenopus
Format Affinity Purified
Gene ID (Entrez) 6656
Conjugate Unlabeled
Host Species Rabbit
Quantity 0.025mg
Immunogen 196 RAVHMkEYPDYkYRPRRk 213
Applications Western Blotting
Regulatory Status RUO
Antigen SOX1