Please login to your online account to display your discounted pricing

anti-SOX1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227189

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227189

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA.
Gene ID (Entrez) 6656
Concentration 0.5mg/mL
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Xenopus
Regulatory Status RUO
Quantity 0.1mg
Primary or Secondary Primary
Conjugate Unlabeled
Test Specificity The amino acidsequence used as immunogen shares significant homology with SOX1, 2, 3, 7, 11, 12, 14, 15, 20, and 21. Dog, Human, New World monkey, Mouse, Opossum, Rat, Xenopus
Immunogen 196 RAVHMkEYPDYkYRPRRk 213
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Antigen SOX1
Format Affinity Purified
Applications Western Blotting