Please login to your online account to display your discounted pricing

anti-Sonic Hedgehog/Shh, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222139SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222139SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 6469
Primary or Secondary Primary
Concentration 1.0mg/mL
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A synthetic peptide made to an N-terminal portion of the human SHH protein (between residues 1-75) [UniProt Q15465]
Formulation PBS
Gene Accession No. Q15465
Conjugate Unlabeled
Host Species Rabbit
Quantity 0.025mL
Format Affinity Purified
Antigen Sonic Hedgehog/Shh