missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Somatostatin Receptor Type 5 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1555650UL
Description
Anti-Somatostatin Receptor Type 5 Antibody is an antibody against Somatostatin Receptor Type 5 for use in western blotting.Specifications
Somatostatin Receptor Type 5 | |
Polyclonal | |
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min). | |
Rabbit | |
Immunoaffinity purified | |
Primary | |
Rat | |
Purified |
Western Blot | |
Unconjugated | |
SSTR5; SS5R | |
Purified peptide from the N-terminus of rat Somatostatin Receptor Type 5 (Accession number P30938). | |
50 μL | |
NM_001053.1 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only