Somatostatin Receptor Type 2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  Emd Millipore Corporation AB9486
Catalog No. AB9486MI
Description
Description
Detect Somatostatin Receptor Type 2 using this Anti-Somatostatin Receptor Type 2 Antibody validated for use in immunohistochemistry (paraffin).
Specifications
Specifications
Somatostatin Receptor Type 2
Unconjugated
Affinity Purified
Rabbit
50μg
Primary
NM_001050.2
Immunohistochemistry (Paraffin)
Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide.
SS2R; SRIF-1
Synthetic peptide from the 2nd extracellular domain of human Somatostatin Receptor Type 2.
−70°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Human