Please login to your online account to display your discounted pricing

anti-Somatostatin R5/SSTR5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056498SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056498T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 6755
Antigen Somatostatin R5/SSTR5
Format Affinity Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. P35346
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A portion of amino acids 300-350 of human SSTR5 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity The Somatostatin Receptor 5 amino acid sequence used as immunogen for this antibody is 100% homologous in human and 94% in chimp. The expected size of SSRT5, based on amino acid content, is approximately 40 kD. It is believed that the 5-10 kD increase in observed size is due to glycosylation. Reacts with Human. Predicted to react with Chimpanzee.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human