Please login to your online account to display your discounted pricing

anti-Somatostatin R5/SSTR5, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human SSTR5 was used as the immunogen.
Antigen Somatostatin R5/SSTR5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056498

 
novus biologicals canada
NB10056498
Unlabeled 0.1mg Each for $255.80

NB10056498T

 
novus biologicals canada
NB10056498SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human SSTR5 was used as the immunogen.
Antigen Somatostatin R5/SSTR5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The Somatostatin Receptor 5 amino acid sequence used as immunogen for this antibody is 100% homologous in human and 94% in chimp. The expected size of SSRT5, based on amino acid content, is approximately 40 kD. It is believed that the 5-10 kD increase in observed size is due to glycosylation. Reacts with Human. Predicted to react with Chimpanzee.
Gene Accession No. P35346
Gene ID (Entrez) 6755
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL