missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Somatostatin R1/SSTR1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide from the C-terminus of rat Somatostatin Receptor 1
Antigen Somatostatin R1/SSTR1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB300120 Novus Biologicals Canada
NB300120
0.1mL Each for $309.05
NB300120SS Novus Biologicals Canada
NB300120SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Somatostatin R1/SSTR1
Antisera
RUO
Specific to Somatostatin Receptor I (SSR1) and does not react with Somatostatin Receptor II (SSR2).
A synthetic peptide from the C-terminus of rat Somatostatin Receptor 1
Polyclonal
Primary
Immunoprecipitation,Western Blot
6751