Please login to your online account to display your discounted pricing

anti-Somatostatin R1/SSTR1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227413

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227413

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 6751
Concentration 0.5mg/mL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mg
Host Species Rabbit
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Somatostatin R1/SSTR1
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Immunogen This antibody was raised against Somatostatin Receptor 1
Format Purified
Applications Western Blotting