missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SODD/BAG4, Polyclonal, Novus Biologicals™

$160.00 - $222.95

Specifications

Host Species Rabbit
Immunogen This antibody was raised against BAG-4
Antigen SODD/BAG4
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227329 Novus Biologicals Canada
NBP227329
0.05mL Each for $222.95
NBP227329SS Novus Biologicals Canada
NBP227329SS
0.025mL Each for $160.00
Specifications Description & Specifications

Specifications

Rabbit
SODD/BAG4
Antisera
RUO
O95429
50μL neat antibody serum.
This antibody was raised against BAG-4
Polyclonal
Primary
Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
9530