Please login to your online account to display your discounted pricing

anti-SODD/BAG4, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 2-16 (KKSGKIKSSYKRE) of human SODD protein were used as the immunogen.
Antigen SODD/BAG4
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056464

 
novus biologicals canada
NB10056464
Unlabeled 0.1mg Each for $255.80

NB10056464T

 
novus biologicals canada
NB10056464SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 2-16 (KKSGKIKSSYKRE) of human SODD protein were used as the immunogen.
Antigen SODD/BAG4
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, bovine and dog, and 93% homologous in rhesus monkey. The 60-65 kD band corresponding to SODD protein is blocked by immunizing peptide. Reacts with Bovine, Dog, Human, Mouse, Rat and Rhesus Monkey. 93% sequence identity with Rhesus monkey protein.
Gene ID (Entrez) 9530
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL