Please login to your online account to display your discounted pricing

anti-SODD/BAG4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056464

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056464

  • / Each
 

Description & Specifications

Specifications

Antigen SODD/BAG4
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS containing 0.2% gelatin
Host Species Rabbit
Immunogen Amino acids 2-16 (KKSGKIKSSYKRE) of human SODD protein were used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9530
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, bovine and dog, and 93% homologous in rhesus monkey. The 60-65 kD band corresponding to SODD protein is blocked by immunizing peptide. Reacts with Bovine, Dog, Human, Mouse, Rat and Rhesus Monkey. 93% sequence identity with Rhesus monkey protein.
Target Species Bovine
Target Species Canine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat