Please login to your online account to display your discounted pricing

anti-SOD1/Cu-Zn SOD, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen The superoxide dismutase enzyme from bovine erythrocytes (CAS No. 9054-89-1) was used as the immunogen.
Antigen SOD1/Cu-Zn SOD
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224915

 
novus biologicals canada
NBP224915
Unlabeled 0.1mg Each for $257.41

NBP224915SS

 
novus biologicals canada
NBP224915SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The superoxide dismutase enzyme from bovine erythrocytes (CAS No. 9054-89-1) was used as the immunogen.
Antigen SOD1/Cu-Zn SOD
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P00441
Gene ID (Entrez) 6647
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL