Please login to your online account to display your discounted pricing

anti-SOCS-5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056639SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056639T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.1mg/mL BSA.
Format Purified
Gene Accession No. O75159
Applications Western Blotting
Immunogen This polyclonal antibody was raised against a synthetic peptide containing amino acid residues of human SOCS5 protein. Human or mouse spleen lysate can be used as a positive control.
Monoclonal or Polyclonal Polyclonal
Antigen SOCS-5
Host Species Rabbit
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9655
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse