Please login to your online account to display your discounted pricing

anti-SOCS-5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056639

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056639

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9655
Formulation PBS containing 0.1mg/mL BSA.
Format Purified
Host Species Rabbit
Gene Accession No. O75159
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Quantity 0.2mL
Immunogen This polyclonal antibody was raised against a synthetic peptide containing amino acid residues of human SOCS5 protein. Human or mouse spleen lysate can be used as a positive control.
Antigen SOCS-5
Applications Western Blotting