Please login to your online account to display your discounted pricing

anti-SOCS-5, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was raised against a synthetic peptide containing amino acid residues of human SOCS5 protein. Human or mouse spleen lysate can be used as a positive control.
Antigen SOCS-5
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056639

 
novus biologicals canada
NB10056639
Unlabeled 0.2mL Each for $255.80

NB10056639T

 
novus biologicals canada
NB10056639SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was raised against a synthetic peptide containing amino acid residues of human SOCS5 protein. Human or mouse spleen lysate can be used as a positive control.
Antigen SOCS-5
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. O75159
Gene ID (Entrez) 9655
Formulation PBS containing 0.1mg/mL BSA.