Please login to your online account to display your discounted pricing

anti-SNX27, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP145282

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP145282

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Conjugate Unlabeled
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 81609
Concentration 1mg/mL
Gene Accession No. Q96L92
Test Specificity Human and mouse. Immunogen sequence has 94% identity to rat and bovine.
Host Species Rabbit
Antigen SNX27
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of human SNX27 (within residues 150-200). [Swiss-Prot No. Q96L92]
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.1mL